missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WoodSafe™ Sharps Bin
Eco-friendly sharps and hazardous waste containers, made from renewable resources using green energy. Reduces CO2 emissions by up to 66%
Brand: WoodSafe™ 1030120
2237.17 SEK valid until 2024-10-18
Use promo code "21642" to get your promotional price.
Alert:
Follow instructions for safe and correct use
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
- Lowers the carbon footprint by >60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
- Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- STOP BURNING PLASTIC!
Specifications
3.3 L | |
Temporary and permanent closure | |
Round | |
0.725 gal | |
16.5 cm X 16.5 cm X 19.8 cm (6.49 x 6.49 x 7.8 in.) | |
50 | |
Yes | |
Autoclavable | |
Safe Disposal of sharps and potentially Infectious Waste or medical waste | |
Biocomposite |
EN ISO 23907-1:2019. UN3291, ADR Approved | |
Brown | |
Environmental friendly Sharpsbin | |
Brown, Yellow | |
Single-use | |
Round | |
Non-sterile | |
Bin, Lid, and Label | |
Cardboard | |
Biohazard Waste Container (Benchtop) |
Lower climate impact