Learn More
Shellac Orange, Pure, Flake, Fisher Chemical™
Product Code. 11488802
Click to view available options
Quantity:
250 g
500 g
Chemical Identifiers
CAS 9000-59-3
MDL Number 148309
Specifications
Packaging Plastic powder jar
Quantity 250 g
Physical Form Solid
Product Identifier
  • Shellac orange