Learn More
Bohlender™ SICCO™ Big Star Desiccator Low
Big Star Desiccator Low made of acrylic glass and aluminum frame.
Brand:  Bohlender™ V1897-07
Product Code. 17117724
  • 16670.00 SEK / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Description
  • Aluminum frame with panels made of acrylic glass
  • Includes one shelf made of stainless steel, tray and hygrometer
  • Usable with up to 5 shelves, stackable
  • Reinforced aluminum frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Non-slip rubber feet
  • Easy to read electronic hygrometer
  • Variable height shelf made of stainless steel
  • Telescopic shelf
  • Desiccant tray
Specifications
Specifications
Acrylic
91 L
550 mm
330 mm
580 mm
13 kg
24 gal.
22 in.
13 in.
23 in.
29 lb.