Learn More
Bohlender™ SICCO™ Mini Desiccator Antistatic Premium
Mini Desiccator Antistatic Premium made of conductive black polycarbonate with bore for earth connection.
Brand: Bohlender™ V1943-06
This item is not returnable.
View return policy
Description
- Black, static dissipative polycarbonate
- Includes two shelves made of static dissipative polyethylene, hygrometer and desiccant
- Non-slip rubber feet, stackable
- All materials used can discharge electrostatic charging by means of a grounding cable which can be connected on the back side (bore diameter 4.3 mm)
- One Touch Door with circular seal
- Robust construction
- Shatterproof cabinet made of polycarbonate
- Hinge door with wide opening angle
- Lightweight, transportable
- Stackable
- Desiccant in a handy, resealable container included in delivery
Specifications
Polycarbonate | |
1.6 gal. | |
8.7 in. | |
7 in. | |
8.4 in. | |
2 lb. |
Silica Gel Jar (23 g) | |
6.2 L | |
221 mm | |
182 mm | |
214 mm | |
0.9 kg |