Learn More
BRAND™ SilberBrand™ Pellet Pattern Automatic Burette
Without intermediate stopcock; with needle-valve stopcock with PTFE spindle
Brand:  BRAND™ 21918
Product Code. 10135880
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Boro 3.3 Glass
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • With needle-valve stopcock with PTFE spindle
  • Schellbach stripe
  • Automatic zeroing
  • Total height, including 2000 mL bottle (soda-lime glass), approximately 1 m.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    50 mL
    0.08 mL
    ±0.08 mL
    Class B
    Borosilicate Glass
    Schellbach Stripe
    Automatic Buret
    DIN EN ISO 385
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products