Learn More
Silica Gel 60 Angstrom Pore Size, 20-45 Micron, for Column Chromatography, Fisher Chemical™
Brand: Fisher Chemical S/0692/60
1145.13 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Specifications
Plastic Powder Jar | |
1 kg |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%