Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
373.95 SEK - 15348.39 SEK
Specifications
Particle Size | -100/200 Mesh |
---|---|
Pore Size | 150 Å |
Packaging | Bottle |
pH | 6.5 to 7.5 |
Surface Area (Metric) | 350 to 400 m2/g |
Type | Silica Gel |
Material | Silica |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375497
|
Thermo Scientific Chemicals
042726.18 |
50 g |
373.95 SEK
50g |
In Stock
Log in to see stock. |
|||||
11385497
|
Thermo Scientific Chemicals
042726.30 |
250 g |
1108.82 SEK
250g |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11395497
|
Thermo Scientific Chemicals
042726.A1 |
1 kg |
3384.46 SEK
1kg |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11365497
|
Thermo Scientific Chemicals
042726.A7 |
5 kg |
15348.39 SEK
5kg |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Description
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
-100/200 Mesh | |
Bottle | |
350 to 400 m2/g | |
Silica |
150 Å | |
6.5 to 7.5 | |
Silica Gel |
RUO – Research Use Only