missing translation for 'onlineSavingsMsg'
Learn More

Silicon Standard For ICP/MS, SPEX CertiPrep™

Product Code. 15275996
missing translation for 'orderingAttributeHoverText'
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Si1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-21-3
Packaging Triple-leached LDPE bottle
Quantity 125 mL
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Si
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES [Si]
IUPAC Name silicon
Molecular Weight (g/mol) 28.09
PubChem CID 5461123
ChEBI CHEBI:27573
Concentration or Composition (by Analyte or Components) 1mg/L
Solvent or Matrix H2O/Tr. F-
Chemical Name or Material Silicon Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'