Learn More
Cytiva Whatman™ O-Ring for use with MV050 Vacuum Filtration Manifolds
Replacement silicone O-ring for use with MV050 Vacuum Filtration Manifolds.
Brand:  Cytiva 10440006
Additional Details : Weight : 0.01100kg
Product Code. 11365144
  • 126.00 SEK / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
  • Product Type: O-Ring
  • Material: Silicone
  • For Use with (Equipment): MV050 Vacuum Filtration Manifold Lids
  • For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Specifications
Specifications
Autoclavable
MV050 Manifold Lids
Non-sterile
MV050/0/09
MV050
Reusable
Silicone
Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
1 Ea.
O-Ring