Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand:  Deutsch Neumann™ 1010556
 View more versions of this product
Product Code. 10588321
  • 154.00 SEK / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
1/Ea.
Transparent
65 mm
Silicone Stopper
200°C
56 mm
Silicone