Learn More
Uvex Silv-Air Premium 5310+ FFP3 Mask
The uvex silv-Air premium product range consists of classic, white flat-fold respirators that fit all kinds of facial shape. With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
Brand: Uvex 8735311
980.63 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
The uvex silv-Air premium+ product line comprises classic folding respirators that adapt to different face shapes which fits perfectly to larger face shapes. The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
- Individually packed
- With exhalation valve
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose and chin area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort
Specifications
P - Oil-Proof | |
Flat-fold | |
FFP3 | |
Particulate Respirator | |
Yes | |
Non-sterile |
White | |
Reusable | |
Polypropylene, Polyester | |
Medium | |
Yes |