missing translation for 'onlineSavingsMsg'
Learn More

SLAMF7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen SLAMF7
Concentration 0.25 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890854
View Documents Promotion Details
Proteintech
12905-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16880854
View Documents Promotion Details
Proteintech
12905-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLAMF7 contains one Ig-like C2-type (immunoglobulin-like) domain. Isoform 1 mediates NK cell activation through a SAP-independent extracellular signal-regulated ERK-mediated pathway. It may play a role in lymphocyte adhesion. Isoform 3 does not mediate any activation. SAP can bind the cytoplasmic tail of isoform 1 when phosphorylated in the presence of Fyn (in vitro). SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Expression was detected in NK cells, activated B-cells, NK-cell line but not in promyelocytic, B-, or T-cell lines. The isoform 3 is expressed at much lower level than isoform 1. There are three named isoforms.
Specifications

Specifications

SLAMF7
Western Blot
Unconjugated
Rabbit
Human
Q9NQ25
57823
SLAMF7 Fusion Protein Ag3804
Primary
-20°C
SLAMF7
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
19A, CD2 subset 1, CD319, CRACC, CS1, Membrane protein FOAP 12, Novel Ly9, Protein 19A, SLAM family member 7, SLAMF7
SLAMF7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications