missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC27A2 Monoclonal Antibody (466106), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524186
This item is not returnable.
View return policy
Description
This antibody stains human FATP2 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
SLC27A2 Monoclonal antibody specifically detects SLC27A2 in Human samples. It is validated for Flow Cytometry.Specifications
SLC27A2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O14975 | |
SLC27A2 | |
HEK293 human embryonic kidney cell line transfected with human FATP2 Met1-Leu620 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
466106 | |
Unconjugated | |
SLC27A2 | |
ACSVL1; Arachidonate--CoA ligase; Facvl1; FATP2; FATP-2; Fatty acid transport protein 2; fatty acid transporter; fatty-acid-coenzyme A ligase, very long-chain 1; hFACVL1; HsT17226; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; SLC27A2; solute carrier family 27 (fatty acid transporter) member 2; solute carrier family 27 (fatty acid transporter), member 2; solute carrier family 27 (fatty acid transporter), member 32; solute carrier family 27 member 2; THCA-CoA ligase; ve; very long-chain acyl-CoA synthetase; very long-chain acyl-Coenzyme A dehydrogenase synthase; very long-chain fatty-acid-coenzyme A ligase 1; very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; Vlac; VLACS; Vlcs | |
Mouse | |
Protein A/G | |
RUO | |
11001 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction