Learn More
Epredia™ Slide File Storage System, blue
Save space by storing materials in the stackable Epredia™ Slide File Storage System.
Brand: Epredia™ B1000700GR
This item is not returnable.
View return policy
Description
Storage:
- Stores 3 × 1″(75 × 26mm) slides
- Stores up to 100 slides (one per slot), 200 slides (two per slot), or 400 slides (slide tray removed)
Specifications
100, 300, 200 | |
400 Slides | |
75 mm | |
26 mm |
Green | |
3 in. | |
1 in. |