Learn More
Hamilton™ Stainless Steel Small Hub Removable Custom Needles
Used for animal injections to limit tissue damage and sample loss.
1825.00 SEK - 3985.00 SEK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
12311583
|
Hamilton™
7804-09 |
Gauge: 23s; Point Style: 2, 3 or 4 |
1825.00 SEK
Pack of 6 |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10509822
|
Hamilton™
207419 |
Gauge: 19; Point Style: 2, 3 or 4 |
1985.00 SEK
Pack of 6 |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12396289
|
Hamilton™
7731-06 |
Gauge: 25s; Point Style: 5 or AS |
3985.00 SEK
Pack of 6 |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12376439
|
Hamilton™
7804-05 |
1825.00 SEK
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12326449
|
Hamilton™
7804-11 |
1825.00 SEK
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12336299
|
Hamilton™
7731-10 |
3985.00 SEK
Pack of 6 |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11512525
|
Hamilton™
207419 |
Gauge: 19; Point Style: 2, 3 or 4 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Autoclavable
- Provided in packs of 6
- Can be used with removable (RN)-style syringes that are 100μL and smaller
- Custom lengths and point styles are available
- Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection