Learn More
Thermo Scientific™ Smart-Vue™ Repeater Modules
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using these repeater modules.
Specifications
Type Repeater Module
Software Type Smart-Vue Client, Smart-Vue Server
Battery Type 3.6 V Lithium
Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Model Repeater Module
View More Specs
Description
Description
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using Thermo Scientific™ Smart-Vue™ Repeater Modules.
Includes:
  • Repeater module
  • Universal (100/240V AC) power supply
  • Wall-mounting holder with velcro
  • Customer reference letter
Specifications
Specifications
Repeater Module
3.6 V Lithium
Repeater Module
Smart-Vue Client, Smart-Vue Server
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.