Learn More
Thermo Scientific™ Smart-Vue™ Repeater Modules
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using these repeater modules.
Specifications
Type | Repeater Module |
---|---|
Software Type | Smart-Vue Client, Smart-Vue Server |
Battery Type | 3.6 V Lithium |
Certifications/Compliance | Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. |
Model | Repeater Module |
Description
Extend the monitoring distance of Thermo Scientific™ Smart-Vue™ Wireless Monitoring Solutions by using Thermo Scientific™ Smart-Vue™ Repeater Modules.
Includes:
- Repeater module
- Universal (100/240V AC) power supply
- Wall-mounting holder with velcro
- Customer reference letter
Specifications
Repeater Module | |
3.6 V Lithium | |
Repeater Module |
Smart-Vue Client, Smart-Vue Server | |
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. |