Learn More
sn-Glycero-3-phosphocholine, MedChemExpress
MedChemExpress sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
Brand: Medchem Express HY-17552
Description
sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.Specifications
sn-Glycero-3-phosphocholine | |
White | |
C8H20NO6P | |
Choline Alfoscerate Alpha-GPC L-α-GPC | |
O=P(OCC[N+](C)(C)C)(OC[C@H](O)CO)[O-] | |
257.22 | |
Research | |
Neuroscience-Neurodegeneration |
28319-77-9 | |
Research | |
10 g | |
H2O : 500 mg/mL (1943.86 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (388.77 mM; Need ultrasonic) | |
257.22 | |
98.0% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only