Learn More
DWK Life Sciences Snap Ring Cap
Snap ring caps provide a convenient solution for closing chromatography vials without the need of crimping or screwing.
Brand:  DWK Life Sciences 225783
337.25 SEK valid until 2025-12-31
Use promo code "24362" to get your promotional price.
 View more versions of this product
Product Code. 16666882
  • 355.00 SEK / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
In contrast to crimp top and screw cap vials, snap ring vials are chosen strictly for convenience. Snap caps may be adequate for applications that do not require heating of the sample above room temperature and where moderate sealing performance can be tolerated.
  • Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red/White/Red
Transparent
Polypropylene, Polyethylene
High Performance Liquid Chromatography (HPLC)
Autoclavable
100
Open Top Cap
11 mm
12.09 mm
PTFE/Silicone/PTFE
45 ± 5 shore A
5.84 mm
1.02 mm
ND11
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Snap Ring Cap > PTFE red, silicone white, PTFE red