missing translation for 'onlineSavingsMsg'
Learn More

SNC 80, Tocris Bioscience™

Product Code. 30001125
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective non-peptide δ agonist

A highly selective and potent non-peptide δ-opioid agonist, 2000-fold selective over μ-opioid receptors.

Chemical Identifiers

CAS 156727-74-1
Molecular Formula C28H39N3O2
Molecular Weight (g/mol) 449.639
InChI Key KQWVAUSXZDRQPZ-UMTXDNHDSA-N
PubChem CID 123924
IUPAC Name 4-[(R)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-(3-methoxyphenyl)methyl]-N,N-diethylbenzamide
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC(=CC=C2)OC)N3CC(N(CC3C)CC=C)C

Specifications

Quantity 50 mg
Synonym snc 80, snc-80, chembl13470, snc80, 4-alpha-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide, dsstox_cid_25662, dsstox_rid_81039, dsstox_gsid_45662, 4-2,5-dimethyl-4-2-propenyl-1-piperazinyl 3-methoxyphenyl methyl-n,n-diethylbenzamide 2s-1 s* ,2alpha,5beta, +-4-alphar-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide
Formula Weight 449.64
Percent Purity >99%
Chemical Name or Material SNC 80
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.