Learn More
Eurofins DiscoverX SNRK Protein, Active
Active, C-terminal 6His-tagged, recombinant, human SNRK, for use in Kinase Assays.
6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 58 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | C-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17300814
![]() |
Eurofins DiscoverX
14-932 |
10 μg |
6046.30 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17310814
![]() |
Eurofins DiscoverX
14-932M |
250 μg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNF-related serine/threonine-protein kinase may play a role in hematopoietic cell proliferation or differentiation. It is a potential mediator of neuronal apoptosis.
GenBank NM_017719
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
C-terminal 6His | |
Truncated | |
SNRK, active |
58 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥74% |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.