Learn More
Jaytec Glass™ Soda Glass Melting Point Tubes
Heat samples in Soda Glass Melting Point Tubes to determine their melting point. These tubes are made of industry-standard soda lime glass.
Brand:  Jaytec Glass™ CAP-MPO-100
 View more versions of this product
Product Code. 12316297
  • 467.00 SEK / Pack of 1000
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Description
  • Designed to be compatible with all major melting point apparatus
  • Available in 75 mm or 100 mm lengths
Specifications
Specifications
Melting Point Instrument
100 mm
Open both ends
1.30 to 1.40 mm
Soda Lime Glass
1000 Pk.