missing translation for 'onlineSavingsMsg'
Learn More
Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals Product Code.: 11491950

Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals

Product Code. 11491950
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11491950

Brand: Thermo Scientific Alfa Aesar A14808.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10580-19-5
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrateShow MoreShow Less
PubChem CID 23666329
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 500 g
Beilstein 4040891
Formula Weight 232.20 (196.17 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium 4-hydroxybenzenesulfonate dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243486 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA14808.22,A14808.0E,A14808.36
Lot NumberD9456A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14808.22,A14808.0E,A14808.36
Lot NumberUSFL002308 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14808.22,A14808.0E,A14808.36
Lot NumberI8837A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14808.22,A14808.0E,A14808.36
Lot NumberUSLF002308 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14808.22,A14808.0E,A14808.36
compliance-icons
Product Identifier
  • Sodium 4-hydroxybenzenesulfonate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium 4-hydroxybenzenesulfonate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.