missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | HDPE plastic bottle |
Safety and Handling
Product Identifier
- Sodium arsenite solution 0.05M (0.1N)
- Danger
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 3
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction