missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium bicarbonate, 99.5%, MP Biomedicals™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium bicarbonate, 99.5%, MP Biomedicals™
Product Code. 11435442 Shop All MP Biomedical Products5 kg
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
This item is not returnable.
View return policy
Product Code. 11435442
Brand: MP Biomedicals 0219549705
This item is not returnable.
View return policy
Description
- It is broadly used as a physiological buffer in cell culture applications.
- The pKa for bicarbonate <-> carbonic acid reaction is 6.4 The pKa for bicarbonate <-> carbonate reaction is 10.3.
- It has two pKas, 6.4 and 10.3. The compound can react with both acids and bases, therefore it can be used as a neutralizer.
- It may be used to remove or wash acidic impurities from a crude liquid to yield a purer sample.
- it reacts with the carboxyl groups to give a fast-forming and effervescent CO2 formation which has been used to test for the presence of carboxylic groups in proteins.

Specifications
Chemical Identifiers
CAS | 144-55-8 |
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Specifications
Color | White |
Physical Form | Powder |
Quantity | 5 kg |
Assay Percent Range | 99.5% |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
Formula Weight | 84.01 |
Percent Purity | 99.5% |
Density | 2.159g/mL |
Chemical Name or Material | Sodium bicarbonate |
Product Suggestions
You may be interested in these alternatives
-
-
-
-
Product Code. 11448856
-
Product Code. 10082840
-
Product Code. 10144713
-
Product Code. 15671310
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium bicarbonate, 99.5%, MP Biomedicals™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction