missing translation for 'onlineSavingsMsg'
Learn More
Sodium Bicarbonate, MP Biomedicals™ Product Code.: 11494632

Sodium Bicarbonate, MP Biomedicals™

Product Code. 11494632 Shop All MP Biomedical Products
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 11494632

Brand: MP Biomedicals 02191435.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used as a neutralizer, since it reacts with both acids and bases. Sodium Bicarbonate, MP Biomedicals is a commonly used laboratory chemical, also referred to as baking soda. It can be an effective way of controlling fungus growth or to remove impur

When reacting with bases, the compound forms carbonates. When reacting with carboxyl groups, it gives a fast-forming and effervescent carbon dioxide formation. Its many uses include:

  • Remove or washes acidic impurities from a crude liquid to yield a purer sample
  • To test for the presence of carboxylic groups in proteins
  • Decreases serum potassium levels, as well as increases blood pH and bicarbonate concentration
  • Decreases the duration of a peripheral nerve block in rats when added to Lidocaine
  • Effective way of controlling fungus growth
  • When mixed with water, can be used as an antacid
  • Used as a leavening agent in cooking and baking (CO2 release causes expansion of the batter and forms the characteristic texture and grain in baked and fried foods)
TRUSTED_SUSTAINABILITY

Specifications

CAS 144-55-8
Physical Form Powder
Quantity 500 g
Molecular Formula CHNaO3
MDL Number MFCD00003528
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
Solubility Information Soluble in water.
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES [Na+].OC([O-])=O
IUPAC Name sodium hydrogen carbonate
Molecular Weight (g/mol) 84.01
PubChem CID 516892
ChEBI CHEBI:32139
Formula Weight 84.01
Percent Purity 99.7 to 100.3%
Grade ACS Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Bicarbonate, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.