missing translation for 'onlineSavingsMsg'
Learn More
Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals Product Code.: 11456282

Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals

Product Code. 11456282 Shop All MP Biomedical Products
100 g
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11456282

Brand: MP Biomedicals 0210289480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Excellent reducing agent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98-99%
Formula Weight 37.8
Chemical Name or Material Sodium Borohydride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.