Learn More
Sodium Caprate, MP Biomedicals
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS | 1002-62-6 |
Molecular Formula | C10H19NaO2 |
MDL Number | MFCD00066453 |
Quantity | 5 g |
Synonym | sodium decanoate, sodium caprate, decanoic acid, sodium salt, decanoic acid sodium salt, sodium caprinate, capric acid sodium salt, sodium-n-decanoate, sodium decanoic acid, capric acid, sodium salt, caprinic acid sodium salt |
InChI Key | FIWQZURFGYXCEO-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCCCCCC([O-])=O |
IUPAC Name | sodium decanoate |
Molecular Weight (g/mol) | 194.25 |
PubChem CID | 4457968 |