missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate, Fisher Chemical™
331.00 SEK - 1540.00 SEK
Chemical Identifiers
CAS | 13472-35-0 |
---|---|
Molecular Formula | H6NaO6P |
Molecular Weight (g/mol) | 156.006 |
MDL Number | 149209 |
InChI Key | VBJGJHBYWREJQD-UHFFFAOYSA-M |
Synonym | sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh |
PubChem CID | 23673460 |
IUPAC Name | sodium;dihydrogen phosphate;dihydrate |
SMILES | O.O.OP(=O)(O)[O-].[Na+] |
Description
Sodium Dihydrogen Orthophosphate, for Electrophoresis, Monobasic, Dihydrate
Chemical Identifiers
13472-35-0 | |
156.006 | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
23673460 | |
O.O.OP(=O)(O)[O-].[Na+] |
H6NaO6P | |
149209 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
sodium;dihydrogen phosphate;dihydrate |
Specifications
60°C | |
White | |
Solid | |
149209 | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate;dihydrate | |
23673460 | |
Plastic powder jar |
13472-35-0 | |
4.5 | |
H6NaO6P | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
O.O.OP(=O)(O)[O-].[Na+] | |
156.006 | |
156.01g/mol |