missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dihydrogen phosphate monohydrate, ExpertQ, Scharlab
Click to view available options
Quantity:
1 kg
Specifications
Specifications
CAS | 10049-21-5 |
Quantity | 1 kg |
Molecular Formula | H5NaO5P |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
SMILES | O.[Na+].OP(O)(O)=O |
IUPAC Name | sodium phosphoric acid hydrate |
Molecular Weight (g/mol) | 139.00 |
Grade | ExpertQ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction