missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sodium Free Salt Mixture for Rat, Modified

Sodium-free salt mixture modified for rat diet.

791.82 SEK - 15855.48 SEK

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
Storage Requirements Room Temperature
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11467311
View Documents
MP Biomedicals™
0296035202
2 kg
791.82 SEK
2kg
Please to purchase this item. Need a web account? Register with us today!
11477311
View Documents
MP Biomedicals™
0296035210
10 kg
3687.74 SEK
10kg
Please to purchase this item. Need a web account? Register with us today!
11487311
View Documents
MP Biomedicals™
0296035250
50 kg
15855.48 SEK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sodium Free Mineral Mixture
  • For rodent diet
  • Powder
Specifications

Specifications

Animal Diet
Rodent diet
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications