missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Concentrate, for 1M solution, Honeywell Fluka™
Click to view available options
Quantity:
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS | 1310-73-2 |
Molecular Formula | HNaO |
Linear Formula | NaOH |
MDL Number | MFCD00003548 |
UN Number | UN1824 |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 6 x 1 Ea. |
SMILES | [OH-].[Na+] |
IUPAC Name | sodium;hydroxide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'