missing translation for 'onlineSavingsMsg'
Learn More
Sodium Metabisulfite, MP Biomedicals™ Product Code.: 11404652

Sodium Metabisulfite, MP Biomedicals™

Product Code. 11404652 Shop All MP Biomedical Products
5 kg
Click to view available options
Quantity:
100 g
5 kg
This item is not returnable. View return policy

Product Code. 11404652

Brand: MP Biomedicals 0219143805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Relied on by scientists. Sodium Metabisulfite, MP Biomedicals is ACS reagent grade with a purity greater than 97%. MP Biomedicals has delivered quality biochemicals for over 50 years.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 5 kg
Formula Weight 190.11g/mol
Percent Purity ≥97.0%
Grade ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Metabisulfite, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.