missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrate, 99+%, ACS reagent, Thermo Scientific™
Click to view available options
Quantity:
500g
1kg
3kg
10kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 10kg |
Assay Percent Range | 99+% |
Linear Formula | NaNO3 |
Fieser | 16,86 |
Merck Index | 15, 8778 |
Solubility Information | Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol |
Formula Weight | 84.99 |
Percent Purity | 99+% |
Grade | ACS Reagent |
Packaging | Plastic drum |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'