missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phenolate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 139-02-6 |
Molecular Formula | C6H5NaO |
MDL Number | MFCD00013134 |
Quantity | 100 g |
Synonym | sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph |
InChI Key | NESLWCLHZZISNB-UHFFFAOYSA-M |
SMILES | [Na+].[O-]C1=CC=CC=C1 |
IUPAC Name | sodium benzenolate |
Molecular Weight (g/mol) | 116.10 |
PubChem CID | 4445035 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'