missing translation for 'onlineSavingsMsg'
Learn More
Sodium Selenite, MP Biomedicals™ Product Code.: 11492021

Sodium Selenite, MP Biomedicals™

Product Code. 11492021 Shop All MP Biomedical Products
10 g
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy

Product Code. 11492021

Brand: MP Biomedicals 0219474110

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sodium Selenite is a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum.

  • Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine.
  • In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides.
  • Sodium selenite is included in various medium supplements for use in cell culture.
  • Sodium selenite has been utilized in studies of cell proliferation and cancer research.
  • It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays.
  • Sodium selenite can inhibit zinc finger protein/DNA interactions.
  • Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
PubChem CID 24934
ChEBI CHEBI:48843

Specifications

Boiling Point Decomposition
Melting Point 1310°F (Decomposition)
Quantity 10 g
SMILES [O-][Se](=O)[O-].[Na+].[Na+]
IUPAC Name disodium;selenite
Molecular Weight (g/mol) 172.948
Formula Weight 172.9
Percent Purity ∼98%
Grade Cell Culture
Chemical Name or Material Sodium Selenite

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Selenite, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.