missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous (White Granules), Fisher BioReagents

Brand:  Fisher BioReagents BP354-500

Product Code. 10285013

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Glass Bottle; 500g

This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium Sulfate Anhydrous
7757-82-6
5 to 8
500 g
Na2O4S
15, 8809
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
0.5% max.
Decomposition Information: >890°C
884°C
White
Solid
≥99.0 %
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99.0%
Glass Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.