missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfide Nonahydrate ACS MP Biomedicals

Product Code. 11427892
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
Synonym Sodium Sulphide, Sodium Sulfide enneahydrate, Disodium Sulfide Nonahydrate
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Quantity 500 g
Formula Weight 240.18
Grade ACS
Density 1.427 at 16 °C (water = 1)
Chemical Name or Material Sodium Sulfide, Nonahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'