missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tetraborate Decahydrate, Reagent Grade, ≥99.5%, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Boiling Point | 1575°C |
Melting Point | 62°C |
Color | Colorless |
pH | 9 to 10 (at 20°C) |
Physical Form | Solid |
Quantity | 5 kg |
Linear Formula | Na2B4O7 · 10H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 381.37g/mol |
Percent Purity | ≥99.5% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'