missing translation for 'onlineSavingsMsg'
Learn More
Sodium Tetrathionate Dihydrate, purum p.a, ≥98.0% (T), Honeywell™ Product Code.: 15648460

Sodium Tetrathionate Dihydrate, purum p.a, ≥98.0% (T), Honeywell™

Product Code. 15648460
25 g, Poly bottle
Click to view available options
Quantity:
25 g
Packaging:
Poly bottle
This item is not returnable. View return policy

Product Code. 15648460

Brand: Honeywell Research Chemicals 72030100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-29-4
Molecular Formula H4Na2O8S4
Molecular Weight (g/mol) 306.24
MDL Number MFCD00150868
InChI Key HAEPBEMBOAIUPN-UHFFFAOYSA-L
Synonym sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2Show MoreShow Less
PubChem CID 203055
IUPAC Name disodium dihydrate (sulfonatodisulfanyl)sulfonate
SMILES O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O

Specifications

Quantity 25 g
Linear Formula NaO3SS2SO3Na · 2H2O
UN Number NONH for all modes of transport
Formula Weight 306.27g/mol
Percent Purity ≥98.0% (T)
Packaging Poly bottle
Chemical Name or Material Sodium tetrathionate dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Tetrathionate Dihydrate, purum p.a, ≥98.0% (T), Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.