missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Vinyl Sulfonate, Monomer, 25% Soln, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 1184-84-5 |
Molecular Formula | C2H3NaO3S |
MDL Number | MFCD09743544 |
Quantity | 100 g |
InChI Key | BWYYYTVSBPRQCN-UHFFFAOYSA-M |
SMILES | [Na+].[O-]S(=O)(=O)C=C |
IUPAC Name | sodium ethenesulfonate |
Molecular Weight (g/mol) | 130.09 |
Formula Weight | 130.109 |
Chemical Name or Material | Sodium Vinyl Sulfonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'