Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
With slim design to allow the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
Brand:  Hamilton™ 55019-17
 View more versions of this product
Product Code. 12382018
  • 2935.00 SEK / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Wide range of volumes available
  • Ejection sleeve made from a high strength, chemically resistant polymer
  • Universal calibration key that reduces the need for costly calibration services
  • Calibration takes only minutes to perform and eliminates downtime
  • Tip nozzle ensures a proper fit with universal pipette tips
  • Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
Specifications
0.40±%
Olive Green
Fixed Volume Pipette
0.18±%
Autoclavable
1
500 μL
1/Pk.