Learn More
Hamilton™ SoftGrip™ Fixed Volume Pipette
With slim design to allow the pipette to reach to the bottom of narrow tubes without the need for extended length tips.
Brand: Hamilton™ 55019-19
Additional Details : Weight : 0.20300kg
Description
- Wide range of volumes available
- Ejection sleeve made from a high strength, chemically resistant polymer
- Universal calibration key that reduces the need for costly calibration services
- Calibration takes only minutes to perform and eliminates downtime
- Tip nozzle ensures a proper fit with universal pipette tips
- Unique color coding and labeling clearly indicates the volume of the pipettes
Specifications
0.30±% | |
Sky Blue | |
Fixed Volume Pipette | |
0.12±% |
Autoclavable | |
1 | |
1000 μL | |
1/Pk. |