Learn More
Thermo Scientific™ Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Brand: Thermo Scientific™
Additional Details : Weight : 2.19000kg
Description
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Maximize capacity on the Thermo Scientific™ Solaris Orbital Shakers with universal platforms and a variety of clamps to fit most vessels.
Certifications
UL, cUL, CE
Services
1 Year
Specifications
Universal Platform | |
12 x 14 in. | |
304.8 mm | |
355.6 mm | |
Solaris 2000 | |
1 Year |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
12 in. | |
14 in. | |
12x14 Universal platform | |
Platform |