Learn More
3M™ Solus™ Anti-Fog Coating
Click to view available options
Lens Tint:
Amber
Brown
Clear
Indoor/Outdoor Gray
Includes:
Foam Gasket
Description
- Adjustable ratchet temples
- Scotchgard™ Anti-Fog Coating resists fogging longer than traditional anti-fog coatings, even after up to 25 washings with water
- 3M internal testing per EN168 test method when compared with traditional anti-fog coatings.
- Indoor/Outdoor Lenses help reduce brightness and glare when working both indoors and outdoors
- Polycarbonate lens absorb 99.9% of UVA and UVB rays
- Meets the requirements of EN166-2001 Standard as an Impact Rated Spectacle
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Indoor/Outdoor Gray |
Frame Color | Gray |
Includes | |
Quantity | 20/Cs. |
Lens Material | Polycarbonate |
Frame Material | Plastic |
Frame Type | Half Frame |