missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Solvex™ Disposable Green Nitrile Chemical Resistant Gloves

Product Code. 12485130
Click to view available options
Size:
7
8
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Solvent resistant, comfortable and protected by fungicidal and bacteriostatic agent. Ansell™ Solvex™ Disposable Green Nitrile Chemical Resistant Gloves are suitable for use in chemical processing and food production.

  • Gauntlet style
  • Disposable
  • Latex free
  • Flocked lining
  • Resistant to chemicals and liquid protection
  • CE, EN374-3:2003, CATEGORY III
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, EN374-3:2003, CATEGORY III
Classification PPE Category III
Coating Material Nitrile
Color Green
Disposable Single-use
Exterior Finish Sandpatch
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 8
Style Gauntlet Cuff
Thickness (Metric) 0.38 mm
For Use With (Application) For chemical processing and preparation, refining - oil & petrol, Metal fabrication, agrochemicals, printing industry
Glove Cuff Style Gauntlet
Liner Material Flocked
Lining Flock Lined
Quantity 12/Pk.
Resistant To Chemicals and liquid protection
Series EDM.37-675
Length (Metric) 330 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'