Learn More
IKA™ SP-Rack
SP-Rack for ROTAVISC spindles offers sufficient space for a variety of different spindles for hanging storage close to the device or for drying.
Brand: IKA™ 0020104809
This item is not returnable.
View return policy
Description
- The stainless steel rack (AISI 304/1.4301)
- For ROTAVISC
- Offers sufficient space for a variety of different spindles for hanging storage close to the device or for drying.
- Dimensions (W x H x D): 290 x 160 x 80 mm
- Weight: 0.633 kg
Specifications
Stainless Steel 1.4301 | |
Hanging Storage for Drying |
Product Content Correction
Product Title
IKA™ SP-Rack > Rack; For Use With: Hanging Storage for Drying
Spot an opportunity for improvement?Share a Content Correction