Learn More
Fisherbrand™ Bonded 1.5mm thick Spacers
For use with notched and plain plates to ensure correct alignment and leak-free casting.
Brand:  Fisherbrand™ VS20S1.5
Product Code. 11854542
  • 212.00 SEK / Pack of 2
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System
Specifications
Specifications
Verti-Gel Maxi 2-Gel Vertical Gel Electrophoresis System
Bonded 1.5 mm thick Spacers