Learn More
Thermo Scientific™ SpeciMAX™ Saliva Collection Kit
The Thermo Scientific SpeciMAX Saliva Collection Kit provides a simple method for the collection of raw (aka plain, whole) saliva.
Brand:  Thermo Scientific™ A50696
 View more versions of this product
Product Code. 17354473
Description
Show More
Specifications
Specifications
Saliva Collection Kit
400 SpeciMAX saliva collection devices (saliva collection tube, cap and funnel)
Store at 15°C to 30°C
Research Use Only
400 Kits
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2209667 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA50696
Lot Number2318875 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA50696
Lot Number2209668 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA50696
Lot Number2309283 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA50696
Lot Number2309283 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog NumberA50696
Product Title
Thermo Scientific™ SpeciMAX™ Saliva Collection Kit > 400 kits