missing translation for 'onlineSavingsMsg'
Learn More

Spectroflux∣r 1000, Lithium tetraborate, fused, <800 micron, Thermo Scientific Chemicals

POA - 2325.00 SEK

Specifications

CAS 12007-60-2
Physical Form Solid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470960
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
096569.A1
1 kg
2325.00 SEK
1kg
View alternative products
Save Up to 
»
Estimated Shipment: 30-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11480960
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
096569.D9
5 x 1 kg
POA
5kg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Often used in testing of cement

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Often used in testing of cement
Specifications

Specifications

12007-60-2
Solid
Videos
SDS
Documents

Documents

Product Certifications