missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards
SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the
QuEChERS analytical techniques (GC, GC/MS, LC/MS)
1155.40 SEK
Specifications
For Use With (Application) | QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) |
---|---|
Volume (Metric) | 5 mL |
Solvents | Acetonitrile |
Product Code | Brand | Concentration | Type | Sorbent | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Type | Sorbent | Price | Quantity & Availability | |||
16329997
|
SPEX CertiPrep™
QUECR-PCB-28 |
50 μg/mL | QuEChERS standard- PCB 28 | PCB 28 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Single Component Reference Standards
- Volume: 5mL
Specifications
QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) | |
5 mL | |
Acetonitrile |